Spex Claritas PPT® Grade ICP-MS Memory Test Solutions
Multi-element standards for ICP-MS analysis
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation.
Use to identify or confirm the maximum concentration of an analyte that does not cause a memory effect greater than the contract required detection limit (CRDL). The memory test solutions are not analyzed directly; equal volumes of the two (Memory Test 1 and 2) are mixed and then introduced into the instrument for a normal sample exposure time. A blank is then run to confirm that all analyte memory effects are below the CRDL.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$303.00 - $544.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Claritas PPT® Grade ICP-MS Memory Test 1 in 5% HNO3; 125 mL
$544.00USD / Each
COMPARE
Multiple 125 Claritas PPT® Grade ICP-MS Memory Test 2 in H2O/tr. HF; 125 mL
$303.00USD / Each