Spex Claritas PPT® Grade Iron for ICP-MS
Single-element standards for ICP-MS analysis
  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
Claritas PPT® grade certified reference materials (CRMs) are designed for inductively coupled plasma (ICP) and ICP-mass spectrometry (MS) analysis. Custom standards can be manufactured upon request.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$57.00 - $127.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
1 µg/mL (1 ppm) 125 Claritas PPT® Grade Iron, 1 µg/mL for ICP-MS in 2% HNO3; 125 mL
$120.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 125 Claritas PPT® Grade Iron, 1,000 µg/mL for ICP-MS in 2% HNO3; 125 mL
$127.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 30 Claritas PPT® Grade Iron, 1,000 µg/mL for ICP-MS in 2% HNO3; 30 mL
$57.00USD / Each
COMPARE
10 µg/mL (10 ppm) 125 Claritas PPT® Grade Iron, 10 µg/mL for ICP-MS in 2% HNO3; 125 mL
$113.00USD / Each