Spex Claritas PPT® Grade Yttrium for ICP-MS
Single-element standards for ICP-MS analysis
  • Made with an acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Certified to DQS to ISO 9001:2015
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
Claritas PPT® grade certified reference materials (CRMs) are designed for inductively coupled plasma (ICP) and ICP-mass spectrometry (MS) analysis. They are made using ultra high purity acids, the highest grade starting materials and high purity water in order to minimize contaminants.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$57.00 - $125.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
1 µg/mL (1 ppm) 125 Claritas PPT® Grade Yttrium, 1 µg/mL for ICP-MS in 2% HNO3; 125 mL
$117.00USD / Each
COMPARE
10 µg/mL (10 ppm) 125 Claritas PPT® Grade Yttrium, 10 µg/mL for ICP-MS in 2% HNO3; 125 mL
$125.00USD / Each
COMPARE
10 µg/mL (10 ppm) 30 Claritas PPT® Grade Yttrium, 10 µg/mL for ICP-MS in 2% HNO3; 30 mL
$57.00USD / Each