Spex Ethanol (Denatured)

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality
  • Designed to work with any instrument and application
Assay percentage: 0.95
Flash point: 14°C
Density: 0.8
Composition: ethyl alcohol 85.40%, water 7%, methyl alcohol 3.60%, methyl isobutyl ketone 1.90%, ethyl acetate 1.30%, petroleum ether 0.80%
Solubility: miscible
Color: colorless liquid
$121.00 - $424.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Form Packaging Volume (mL)
Availability Pricing
COMPARE
Liquid Natural poly bottle 4000
$121.00USD / Each
COMPARE
Liquid Round metal can 20000
$424.50USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat