Spex ICP-MS Interferents B3 Standards, 2 µg/mL
Multi-element standards for ICP-MS instrumentation
  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • CLP ISM 01.2 and ILM 05.2 & 05.3 Standards for ICP-MS
Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.
US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentration levels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements
For verification of inter-element and background correction factors at the beginning and the end of each analysis run. In addition, a verification must be done after every 20th sample. Two solutions are required for the most common interference check: Solution A, Interferents A2 (04569-10) and Solution AB, a combination of Interferents A2 (04569-10) and analytes (04569-11 or 04569-13). Solution A is prepared by diluting 04569-10 ten-fold. Solution AB is prepared by diluting 04569-10 ten-fold and 04569-11 or 04569-13 hundred-fold; for example, 10 mL of 04569-10 and 1 mL of 04569-11 or 04569-13 into a 100 mL volumetric flask, brought to volume with a matrix blank. Once prepared, the solutions should be analyzed consecutively, starting with Solution A.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$400.00 - $418.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
125 ICP-MS Interferents B3 Set with Mercury, 2 µg/mL; 125 mL
$418.50USD / Each
COMPARE
125 ICP-MS Interferents B3 without Mercury, 2 µg/mL in 2% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
$400.00USD / Each