Spex Multi-Element Biodiesel Standards, 5 Elements
Standards specifically designed for industrial use
  • Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025
  • Meet the requirements for testing ASTM Methods D6751, D5453 and EN 14216
Governments worldwide have passed regulations that mandate lower levels of sulfur in biodiesel fuel. To comply with the implementation of these regulations, SPEX CertiPrep offers specifically designed Certified Reference Materials for industrial use. Our B100 Biodiesel Standards meet the requirements for testing ASTM Methods D6751, D5453 and EN 14214.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$234.00 - $246.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
5 µg/g (5 ppm) 100 Biodiesel Standard, 5 Elements, 5 µg/g in B100 Biofuel; 100 mL
$238.00USD / Each
COMPARE
10 µg/g (10 ppm) 100 Biodiesel Standard, 5 Elements, 10 µg/g in B100 Biofuel; 100 mL
$246.00USD / Each
COMPARE
20 µg/g (20 ppm) 100 Biodiesel Standard, 5 Elements, 20 µg/g in B100 Biofuel; 100 mL
$234.00USD / Each