Spex USP <232> Revision 40, Oral 3 Elemental Impurities Mixes
Use with ICP-MS instrumentation for the analysis of low level impurities
  • Accredited by A2LA to ISO/IEC 17025:2017 & ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Helps meet the new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury
The new guidelines set by the United States Pharmacopeia (USP) have pushed the pharmaceutical and nutraceutical industries to provide accurate, quantifiable results for metal analysis in drugs, pharmaceutical substances and raw materials.
USP <232> outlines new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury. ICP-MS instrumentation, along with accurate ICP-MS standards, allow for increased efficiency and accuracy of the analysis necessary to comply with the new regulations.
Use these standards can be used as a calibration or check standard to verify Oral Daily Dose PDE, Parenteral Component Limit or Parenteral Daily Dose PDE as well as Inhalation Component Limit or Daily Dose.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$231.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Matrix Volume (mL) Description
Availability Pricing
COMPARE
10% HNO3 125 USP <232> Revision 40, Oral 3 Mix 1 Elemental Impurities in 10% HNO3; 125 mL
$231.00USD / Each
COMPARE
5% HNO3/tr. Tartaric Acid/tr. HF 125 USP <232> Revision 40, Oral 3 Mix 2 Elemental Impurities in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
$231.00USD / Each