Taylor 6092N Series Food Service Bimetal Pocket Thermometers
Color Coded Thermometers Food Themometers
  • 1" Diam. Dial and 5" stem
  • Shatter-proof lens and pocketed sleeve
  • 0°F to +220°F temperature range
  • NSF listed
  • Stainless steel construction
  • Five colors to optimal food safety
Color coded food safety pocket thermometer. This Taylor Precision 6092N, also known as a temperature indicator, is constructed of stainless steel. It has a 1" diameter dial and a 5" stem. This shatter-proof Taylor Precision 6092N has a pocketed sleeve. The sleeve features a unique built-calibration feature. This thermometer, Taylor Precision 6092N reads temperatures ranging from 0°F to +220°F. It is NSF listed. This Taylor Precision 6092N is available in five colors. Kitchens can use these pocket thermometers to color code. This food safe Taylor Precision 6092N helps to prevent cross-contamination. Purple is ideal for allergens, yellow for poultry, red for raw meat, brown for cooked meat and blue for fish.
ITEM(S) ON SALE
$8.96USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
Taylor 6092NBL Color-Coded Cross-Contamination Bimetal Pocket Thermometer - Blue EW-90025-70 Mfr # 6092NBLBC
COMPARE
220 0 Cross Contamination 1" Bimetalal Pocket Thermometer - Blue
$8.96USD / Each
$11.20USD / Each