Tegam Datalogging Thermocouple Thermometers with Wireless Connectivity
Collect an unlimited number of time-stamped measurements using wireless connection
  • Stores up to 1000 measurements on the meter
  • ±(0.04% of reading + 0.3°C) accuracy
  • Accepts thermocouple types J, K, T, E, N, R, S, and B
  • Ergonomically designed to minimize user fatigue
  • Easy-to-clean seamless case
Easily collect, store, and transmit temperature data for analysis or compliance reporting with these datalogging thermometers. Meters accurately record and store up to 1000 time-stamped measurements at the touch of a button on the meter; collect an unlimited amount of measurements by transferring them to a phone, PC, or cloud storage using Bluetooth® wireless technology. Simply download the Thermomoter Link™ mobile app for a two-way communication and control.
Both 931B and 932B models accept type J, K, T, E, N, R, S, and B thermocouple probes and have accuracy of ±(0.04% of reading + 0.3°C). Use the probe offset feature to zero out temperature probe errors, significantly improving overall measurement uncertainty. Increase productivity, efficiency, and quality using real-time statistical data. Thermometers display min, max, average, range, and standard deviation on demand, providing critical information and helping to analyze manufacturing process to make immediate adjustment for maximum efficiency and quality. Press the HOLD button to freeze display for reading later.
Easily view the readings on a large backlit LCD. The ergonomically designed thermometer allows for one-handed operation and minimizes user fatigue during extended use. The smooth, seamless case resists dirt and dust and is easy to clean. Thermometers provide up to 1000 hours of battery life using three AA batteries. Available in single- or dual-input model.
$511.00 - $573.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Logging Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
Tegam 931B Datalogging Thermocouple Thermometer with Wireless Connectivity, Single-Input EW-78909-34 Mfr # 931B
COMPARE
Yes 1 Types J, Type K, Type T, Type E, Type B, Type N, Type R, Type S
$511.00USD / Each
Tegam 932B Datalogging Thermocouple Thermometer with Wireless Connectivity, Dual-Input EW-78909-35 Mfr # 932B
COMPARE
Yes 2 Types J, Type K, Type T, Type E, Type B, Type N, Type R, Type S
$573.00USD / Each