Testo 103 / 104 Folding Food Thermometers
Compact, rugged design for everyday use!
  • Hinged design conveniently folds to fit in your pocket
  • Narrow probe tip for optimum measuring flexibility in the kitchen and at the counter.
  • ABS housing is easy to keep clean.
  • Rugged (IP55) protection for daily use and safe cleaning.
Use the testo folding food thermometers in the kitchen for quick and convenient spot-checks on all types of food....just unfold and start measuring. Get instant feedback on the large LCD display in seconds in °C or °F from -58 to 482°F depending upon model. Both models feature an NTC sensor , auto off and 0.9°F accuracy.
$122.00 - $196.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Min Temperature (° F)
Availability Pricing
Testo 0560 0103 Compact Folding Food Thermometer, Narrow; -22 to 428 F EW-90025-22 Mfr # 0560 0103
COMPARE
428 Digital Indicator -22
$122.00USD / Each
Testo 0563 0104 Waterproof Folding Food Thermometer, Narrow; -58 to 482 F EW-90025-23 Mfr # 0563 0104 01
COMPARE
482 Digital Indicator -58
$196.00USD / Each