Testo 108 Food Thermometer Accessory Probes
Accurate Type T thermocouple probes idea for food applications
The Testo 0628 0027 waterproof Type T thermocouple needle probe measures temperature from -50 to 250 degrees C (-58 to 482 degrees F). Probe measures 150mm (5.9") and has a 1.4mm (0.05") diameter tip for reduced impact and features a response time of 2 seconds for fast readings.
The Testo 0603 3392 IP67 waterfproof food probe measures temperature from -50 to 350 degrees C (-58 to 662 degrees F). Probe measures 4.9" long and has a 0.16" diameter. This probe features a response tome of 7 seconds.
The Testo 0603 2192 IP67 waterfproof stainless steel probe measures temperature from -50 to 350 degrees C (-58 to 662 degrees F). Probe measures 4.9" long and has a 0.16" diameter and comes with a PUR cable. This probe features a response tome of 7 seconds.
$167.00 - $230.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Description
Availability Pricing
Testo 0628 0027 Type T Thermocouple Waterproof Needle Tip Food Probe, -58 to 482FIP-65 Rated EW-90025-26 Mfr # 0628 0027
COMPARE
482 Thermocouple Type T Thermocouple Waterproof Needle Tip Food Probe, -58 to 482FIP-65 Rated
$230.00USD / Each
Testo 0603 2292 Thermocouple Food Probe, Type T, SS, Waterproof EW-90025-28 Mfr # 0603 2192
COMPARE
662 Thermocouple Thermocouple Food Probe, Type T, Ss, Waterproof
$167.00USD / Each