Testo Temperature Thermistor Probes

Different probe styles accommodate a variety of applications
Thermistors exhibit a greater senstitivity and accuracy in the biological range. Probes encase a ceramic element that generally decreases in resistance as the temperature increases.
$129.00 - $145.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Testo 0613 1712 Thermistor Air Probe -58 to 300°F (-50 to 148°C) EW-37803-56 Mfr # 0613 1712
COMPARE
Thermistor Air Probe -58 to 300°F (-50 to 148°C)
$129.00USD / Each
Testo 06131912 Thermistor Surface Probe -58 to 300°F (-50 to 148°C) EW-37803-57 Mfr # 0613 1912
COMPARE
Thermistor Surface Probe -58 to 300°F (-50 to 148°C)
$145.00USD / Each
Testo 0613 1212 Thermistor Penetration Probe -58 to 300°F (-50 to 148°C) EW-37803-58 Mfr # 0613 1212
COMPARE
Thermistor Penetration Probe -58 to 300°F (-50 to 148°C)
$129.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat