Thermo Scientific Dual Star pH/ISE Meter Kits
No manual needed to begin measuring
  • Dual inputs allow you to simultaneously measure and display pH, ISE and temperature
  • Simple to operate — prompts to walk you through each meter operation
  • More options for ISE than any other meter
The Thermo Scientific Orion Dual Star meter provides dual BNC inputs for simultaneous pH and ISE readings. Ion determination is facilitated by additional options for concentration units, optional auto-blank for low-level measurements, built-in Star-stirrer control, and the ability to view and print your calibration graph with slope data. The meter easily guides you through common direct measurements, but also is programmed for a range of Known Addition and Subtraction methods. Autosampler capabilities allow you to take advantage high sample throughput technology to save time and improve measurement quality.
The Dual Star meter uses Smart Stability™ and Averaging™ functions to ensure accurate results. The meter offers a full complement of GLP-related features including 1000 point data logging with time and date stamp, electrode verification, and full calibration data. Generate and store up to 10 password protected methods per channel. Easily download data from the USB port or bidirectional RS-232.
$3,915.00 - $4,420.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max ISE Max pH (pH) Description
Availability Pricing
Thermo Scientific Dual Star Meter Ammonia Kit EW-58825-11 Mfr # 2115201
COMPARE
19999 ppm 20 Meter Ammonia Kit
$4,140.00USD / Each
Thermo Scientific Dual Star Meter Chloride Kit EW-58825-12 Mfr # 2115202
COMPARE
19999 ppm 20 Meter Chloride Kit
$4,300.00USD / Each
Thermo Scientific Dual Star Meter Fluoride Kit EW-58825-13 Mfr # 2115203
COMPARE
19999 ppm 20 Meter Fluoride Kit
$4,420.00USD / Each
Thermo Scientific Dual Star Meter Sodium Kit EW-58825-15 Mfr # 2115205
COMPARE
19999 ppm 20 Meter Sodium Kit
$3,915.00USD / Each