Thermo Scientific Heratherm Advanced Protocol Incubator
Unique dual convection technology allows excellent temperature performance, providing an optimal sample environment!
  • Internal socket allows the use of other lab equipment inside the unit/
  • Microprocessor control with vacuum fluorescent display
Broad temperature range from 5°C above ambient to 105°C is suitable for drying applications. Dual convection allows for different applications ; fan speed is adjustable from 0 to 100%. Corrosion-resistant stainless-steel interior is easy to clean and internal glass door allows sample viewing without impacting temperature. Digital timer can be set for daily or weekly On/OFF cycles. Small footprint saves space in your lab.
$4,400.00 - $6,590.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm 51028066 Advanced Protocol 2 cuft Dual Convection Incubator, 120V EW-38800-08 Mfr # 51028066
COMPARE
2.3 105 120
$4,400.00USD / Each
Thermo Scientific Heratherm 51028068 Advanced Incubator, 6.4 cu ft, Dual; 120V EW-38800-12 Mfr # 51028068
COMPARE
6.3 105 120
$6,590.00USD / Each