Thermo Scientific Nalgene Erlenmeyer Flasks, Polycarbonate
Graduated polycarbonate (PC) flasks include polypropylene screw closures. Flasks have excellent clarity and good physical strength. Use PC flasks for many culturing techniques. Limited autoclavability*.
$149.00 - $214.00USD / Pkg of 6
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Cap Type
Availability Pricing
Thermo Scientific Nalgene 4108-0125 PC Screw Top Erlenmeyer Flask, 125 mL, 6/Pk
Buy More Save More
EW-06112-52 Mfr # 4108-0125
COMPARE
112 125 33 mm thread
$159.00USD / Pkg of 6
Thermo Scientific Nalgene 4108-0250 PC Screw Top Erlenmeyer Flask, 250 mL, 4/Pk
Buy More Save More
EW-06112-53 Mfr # 4108-0250
COMPARE
138 250 38 mm thread
$149.00USD / Pkg of 4
Thermo Scientific Nalgene 4108-0500 PC Screw Top Erlenmeyer Flask, 500 mL, 4/Pk
Buy More Save More
EW-06112-54 Mfr # 4108-0500
COMPARE
178 500 43 mm thread
$214.00USD / Pkg of 4