Thermo Scientific Nalgene Polycarbonate Storage Boxes
Storage boxes can withstand temperatures down to -196°C
Storage boxes 06754-57 and -59 hold microcentrifuge tubes. Box 06754-58 holds bags, ampules, bottles, and other containers; withstand temperatures down to -196ºC. Clear top is keyed for easy sample identification. Boxes fit Nalgene CryoBox™ Rack 06756-08, and other standard stainless steel cryogenic storage racks. Autoclavable.
$112.00 - $142.00USD / Pkg of 4
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Tube Size Description
Availability Pricing
Thermo Scientific Nalgene 5055-5005 PC Storage Box, 81 x 0.5 mL Vials; 4/Pk EW-06754-57 Mfr # 5055-5005
COMPARE
0.5 mL PC Storage Box, 81 x 0.5 mL Vials; 4/Pk
$141.00USD / Pkg of 4
Thermo Scientific Nalgene 5050-0001 Polycarbonate storage boxes, 4/pack EW-06754-58 Mfr # 5050-0001
COMPARE
Not divided Polycarbonate storage boxes, 4/pack
$112.00USD / Pkg of 4
Thermo Scientific Nalgene 5055-5015 PC Storage Box, 64 x 1.5 mL Vials; 4/Pk EW-06754-59 Mfr # 5055-5015
COMPARE
1.5 mL PC Storage Box, 64 x 1.5 mL Vials; 4/Pk
$142.00USD / Pkg of 4