Thermo Scientific Nalgene Unwire Half Racks, Resmer
Half racks save space—they’re half as long as full-size Unwire racks with the same durable, polyketone (PK) construction. Choose from four test tube sizes. Autoclavable.
$43.30 - $49.50USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Min Tube Size (mm) Tubes Held
Availability Pricing
Thermo Scientific Nalgene 5972-0513 Unwire half rack for 13-mm tubes, red
Buy More Save More
EW-06738-50 Mfr # 5972-0513
COMPARE
Red 13 36
$43.30USD / Each
Thermo Scientific Nalgene 5972-0013 Unwire half rack for 13-mm tubes, white
Buy More Save More
EW-06738-55 Mfr # 5972-0013
COMPARE
White 13 36
$44.30USD / Each
Thermo Scientific Nalgene 5972-0516 Unwire half rack for 16-mm tubes, red
Buy More Save More
EW-06738-60 Mfr # 5972-0516
COMPARE
Red 16 36
$45.30USD / Each
Thermo Scientific Nalgene 5972-0016 Unwire half rack for 16-mm tubes, white
Buy More Save More
EW-06738-65 Mfr # 5972-0016
COMPARE
White 16 36
$45.30USD / Each
Thermo Scientific Nalgene 5972-0020 Unwire half rack for 20-mm tubes, white
Buy More Save More
EW-06738-75 Mfr # 5972-0020
COMPARE
White 20 20
$49.50USD / Each
Thermo Scientific Nalgene 5972-0025 Unwire half rack for 25-mm tubes, white
Buy More Save More
EW-06738-80 Mfr # 5972-0025
COMPARE
White 25 25
$49.50USD / Each
Thermo Scientific Nalgene 5972-0325 Unwire half rack for 25-mm tubes, blue
Buy More Save More
EW-06738-83 Mfr # 5972-0325
COMPARE
Blue 25 25
$49.50USD / Each
Showing 1- 7 of 7