Thermo Scientific Orion Star A 300 Conductivity and pH/Conductivity Meters
Advanced features and dependability makes it the top choice for measuring samples
  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A322 conductivity and A325 pH/conductivity meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only and as complete meter kit configurations
$1,480.50 - $3,846.15USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A322 Star A322 Conductivity Portable Meter Only EW-58825-56 Mfr # STARA3220
COMPARE
Star A322 Conductivity Portable Meter Only
$1,480.50USD / Each
Thermo Scientific STAR A322 Star A322 Conductivity Portable Meter Kit EW-58825-58 Mfr # STARA3225
COMPARE
Star A322 Conductivity Portable Meter Kit
$2,110.50USD / Each
Thermo Scientific STAR A325 Star A325 pH/Cond Portable Meter Kit EW-58825-70 Mfr # STARA3255
COMPARE
Star A325 pH/Cond Portable Meter Kit
$3,846.15USD / Each