Thermo Scientific Orion Star A200 DO/RDO/BOD/Temperature Benchtop Meter
Durability and dependability make it the ideal choice for every lab, every sample, every day
  • Multlanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Choose between RDO optical probe and polarographic probe
  • Directly measure in mg/L (ppm) or % saturation
  • Automatic temperature, barometric pressure abd salinity corrections
  • Nonvolatile memory holds up to 200 data points with time date stamp
  • Individual passwords provide better method security
  • Option of AC power or four AA batteries (sold separately)
Thermo Scientific Orion Star A213 dissolved oxygen meters offer the option of RDO optical probes or traditional polarographic probes. Meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD shows measurement and temperatrure along with electrode ststus, time and date stamp, sample ID, user ID and calibration data.
GLP related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only, DO or BOD kit configurations.
$3,278.10 - $3,747.45USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A213 Star A213 DO Benchtop Meter Kit EW-53101-04 Mfr # STARA2135
COMPARE
Star A213 DO Benchtop Meter Kit
$3,278.10USD / Each
Thermo Scientific STAR A213 Star A213 DO/BOD Benchtop Meter Kit EW-53101-06 Mfr # STARA2136
COMPARE
Star A213 DO/BOD Benchtop Meter Kit
$3,747.45USD / Each