Thermo Scientific Precision Shaking Water Baths

Easily clean and maintain bath with coil-free design
  • Oscillations per Min: 0, 30-200
  • Temperature range: 5 C above ambient to 100 C
  • Temperature uniformity at 70 C: ± 0.05 C
  • Temperture stability at 70 C: ± 0.1 C
Thermo Scientific Precision shaking water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. Precision shaking water baths are available in 15 L and 27 L capacities.

Precision shaking water baths are available in 15 L and 27 L capacities. The shallow form model has a capacity of 15 L with a removable tray that provides a depth of 3.5” (8.9 cm) for use with smaller sample containers. The Dubnoff water bath is designed specifically for applications that require your samples to be incubated in a controlled atmosphere; equipped with one large and two small gassing hoods.
$4,140.00USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Reservoir Capacity (L) Description
Availability Pricing
Thermo Scientific Precision TSSWB15S Shaking Water Bath, +/-0.1C, 15L Shallow EW-12150-00 Mfr # TSSWB15
COMPARE
15 Shaking Water Bath, +/-0.1C, 15L
$4,140.00USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat