Thermo Scientific™ SuperNuova+™ Digital Stirrers and Stirring Hot Plates
Advanced features and user-friendly controls maximize performance
  • Programmable set points for time, rpm, and heat easily link together to create ramp and dwell protocols
  • StirTrac™ function provides consistent slow speed stirring, precise speed control, and stong magnetic coupling
  • External 100 Pt RTD probe, along with field calibration, provides accurate sample temperature feedback
  • Built-in timer turns off heating, stirring, or both operations
  • Multiline LED display shows current and set point temperatures simultaneously
  • Hot top warning system prevents accidental burns when heating surface is above 50°C (hot plate functionality only)
  • New design features a large two-line LED display raised to protect from splashes and spills
The SuperNuova+ stirrers and stirring hot plates incorporate additional safety features to protect samples. The over-temperature protection is adjustable, cutting power in the event of a heater malfunction. A configurable temperature limit setting provides an additional layer of safety. Optional 1000 Pt RTD enhances temperature feedback accuracy.
Newly designed display is tilted to be easily read from above. To prevent damage from splattered liquids, the display is raised, diverting spills away from electronics.
$1,490.00 - $2,320.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in) Top Plate Material
Availability Pricing
Thermo Scientific Supernuova S88857190 Stirrer, 7x7" Ceramic; 120 VAC EW-04600-27 Mfr # S88857190
COMPARE
120 7 1/4 Ceramic
$1,490.00USD / Each
Thermo Scientific Supernuova SP88857190 Stirring Hot Plate, 7x7" Ceramic; 120 VAC EW-04600-29 Mfr # SP88857190
COMPARE
120 7 1/4 Ceramic
$1,670.00USD / Each
Thermo Scientific Supernuova SP88857195 Stirring Hot Plate, 7x7" Ceramic; 230 VAC EW-04600-31 Mfr # SP88857195
COMPARE
230 7 1/4 Ceramic
$1,700.00USD / Each
Thermo Scientific Supernuova SP88857198 Stirring Hot Plate, 7x7" Aluminum; 230 VAC EW-04600-35 Mfr # SP88857198
COMPARE
230 7 1/4 Aluminum
$2,030.00USD / Each
Thermo Scientific Supernuova S88850190 Stirrer, 10x10" Ceramic; 120 VAC EW-04600-55 Mfr # S88850190
COMPARE
120 10 1/4 Ceramic
$1,870.00USD / Each
Thermo Scientific Supernuova SP88850190 Stirring Hot Plate, 10x10" Ceramic; 120 VAC EW-04600-60 Mfr # SP88850190
COMPARE
120 10 1/4 Ceramic
$2,320.00USD / Each