Traceable® Digital Benchtop Radio-Controlled Atomic Clocks
Extremely accurate—to 1/10,000 of a second!
  • Clear display with elegant acrylic lens shows a resolution of 1 second
  • Displays time, calendar, and weekday
  • Reads indoor temperature (°C/°F)
  • Offers alarm and snooze function
  • Use an amber LED backlight for daytime use and an auto-night light function for nighttime
These digital wall clocks contain a pre-tuned internal radio receiver that receives a radio frequency signal generated by the U.S. Atomic Clock in Boulder, Colorado. With an accuracy of 1/10,000 of a second, the clocks automatically adjust to precise time (12 or 24-hr format), daylight savings time, month, date, and day of the week. Also view the ambient temperature, signal strength, and time zone map on the LCD.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$33.20 - $49.70USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Traceable Mini Digital Benchtop Atomic Clock
Always in Stock
EW-62002-08 Mfr # 1065TR
COMPARE
Mini Digital Benchtop Atomic Clock
$33.20USD / Each
Traceable Compact Digital Benchtop Atomic Clock
Always in Stock
EW-62002-09 Mfr # 1066TR
COMPARE
Compact DIgital Benchtop Atomic Clock
$49.70USD / Each