Traceable® Digital Thermometers with Memory Card and Calibration
Captures up to 5.9 million readings on a removable SD card
  • Set measurement intervals from once per minute up to once every 720 minutes
  • Meets all current CDC requirements and recommendations for vaccine monitoring and storage
  • Text report includes date/time of day and sensor readings
  • Audible alarm and LED flashes when temperature exceeds alarm set points
  • Simultaneously view probe temperature, ambient temperature, humidity, and time
These thermometers record data onto a standard SD memory card instead of built-in memory—simply exchanged out card when full for unlimited data storage. Send text report including date, time of day, and readings of sensors to any spreadsheet or database. A memory card reader (included) plugs into any USB port to provide an instant report.
The unique 4-line display simultaneously shows external probe temperature, ambient temperature, internal humidity, and time of day. An audible and visual alarm alerts you when temperature levels are higher or lower than the programmable set points. User-settable parameters include date, time, sampling time, temperature unit of measure, high/low alarm set points, and auto power off.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$409.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Temperature Probe Type
Availability Pricing
Traceable Digital Thermometer with Memory Card; 1 Bottle Probe EW-37803-84 Mfr # 6400TR
COMPARE
Bottle
$409.00USD / Each
Traceable Digital Thermometer with Memory Card; 1 Stainless Steel Probe
Always in Stock
EW-37803-85 Mfr # 6401TR
COMPARE
Stainless Steel Immersion
$409.00USD / Each