Traceable® Infrared Thermometers
Safely and easily locate hot and cold spots, preventing problems and saving costs
  • Pistol-grip handle is comfortable and easy to use
  • Noncontact infrared thermometers deliver accurate temperature readings from a distance
  • Trigger lock for continuous scanning
Infrared thermometers have become the preferred tool for diagnostics and inspection where temperature is an indicator. Traceable infrared thermometers are valued by professionals in a variety of industries—from industrial maintenance to foodservice—because they quickly, accurately, and safely measure surface temperature. Plus, the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
Since infrared (IR) meters do not contact the objects being measured, they are the safest way to measure hot, hard-to-reach, or moving parts while eliminating potential damage and contamination. IR thermometers are easy to use: just pull the trigger and the temperature will show in an instant on a large, backlit display.
All models feature Min/Max and Hold functions, °F/°C selectable units, single targeting laser, large backlight LCD, and auto power-off. Models 20250-05 and -06 also offer adjustable emissivity and user-adjustable temperature alarm with indicator light.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$142.00 - $176.25USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
Traceable Infrared Thermometer with Fixed Emissivity, 10:1
Always in Stock
EW-20250-04 Mfr # 9004TR
COMPARE
10:1 Fixed at 0.95 630
$142.00USD / Each
Traceable Infrared Thermometer with Adjustable Emissivity and Alarm, 12:1
Always in Stock
EW-20250-05 Mfr # 9005TR
COMPARE
12:1 Adjustable from 0.10 to 1.0 1000
$176.25USD / Each
COMPARE
20:1 Adjustable from 0.10 to 1.0 1472
$152.75USD / Each