Traceable® Jumbo-Display Dial Thermometers with Calibration
Easily view temperature reading on the large display
  • Display updates every second
  • Smooth, stainless steel probe is perfect for sticky and viscous solutions
  • Readings update every second
  • °C/°F selectable
Large dial head with 3/8" high, bright LCD is easy to read, even from a distance. Wide temperature range of -50 to 150°C (-58 to 302°F) covers most laboratory temperature measurement needs. Stainless steel probe and high-impact, chemical-resistant case offer durability.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$45.10 - $76.20USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Accuracy Probe Type
Availability Pricing
Traceable Jumbo-Display Dial Thermometer, ±1°C Accuracy; 1 Flat-Surface Probe
Buy More Save More
EW-36842-95 Mfr # 4355TR
COMPARE
±1°C (-20 and 100°C) Flat surface
$67.50USD / Each
Traceable Jumbo-Display Dial Thermometer, ±1°C Accuracy; 1 Piercing-Tip Probe
Always in Stock
EW-98767-22 Mfr # 4049TR
COMPARE
±1°C (-20 and 100°C) Piercing tip
$45.10USD / Each
Traceable Jumbo-Display Dial Thermometer, ±0.3°C Accuracy at Tested Points; 1 Piercing-Tip Probe
Always in Stock
EW-98767-41 Mfr # 4374TR
COMPARE
±0.3°C at tested points; otherwise ±1.0°C Piercing tip
$76.20USD / Each