Traceable® Pressure Gauges with Calibration
Displays pressure in eight different units of measure
  • Zero the unit at the touch of a button
  • Serial output port allows connection to a PC
  • Features Min, Max, and Hold readings
Measure pressure and view in one of eight different units of measure (psi, bar, m/H2O, atmospheres, mm/Hg, inch/Hg, inch/H2O, or kg/cm2) with these pressure gauges. Simply press the HOLD button to freeze the reading; 3/8"H digits are easy to read. Connect to a PC using the serial interface. A data acquisition software 37804-13 (sold separately) makes it easy to store and analyze the captured data. The included transducer has a 1/4" NPT(M) connection.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$669.00 - $681.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Resolution
Availability Pricing
Traceable Pressure/Vacuum Gauge; -14.7 to 725 psi
Always in Stock
EW-98766-89 Mfr # 3165TR
COMPARE
725 1 psi
$681.00USD / Each
Traceable Pressure/Vacuum Gauge; -14.7 to 29.0 psi
Always in Stock
EW-98766-90 Mfr # 3166TR
COMPARE
29 0.02 psi
$669.00USD / Each