Traceable® Two-Channel Digital Thermometers with Calibration
Monitor two areas at the same time
  • Displays date and time when Min/Max temperatures occur
  • Audible and visual alarms alert you when alarm conditions occur
  • 3-1/2 ft long micro cable permits a door to close on it
Use the dual-channel thermometers for monitoring temperature in two separate areas or measuring two adjoined areas to ensure uniformity. Two channels record the date and time of the minimum and maximum readings, while independently monitoring user programmable high and low alarms. Set alarm in 1° increments to provide an alert when temperature readings exceed the preset alarm limits. Once alarm is triggered, an audible alarm sounds and an LED flashes; if the readings return to acceptable limits, the audible alarm will turn off, however, the LED will continue to flash until reset.
Probe is attached to a 3-1/2-ft (1.05 m) micro-cable that permits a door to close on it; probe and cable are both submersible. Mount meter with included hook-and-loop fastener, magnetic strips, or use flip-open stand on benchtop.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$54.40 - $90.04USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Probe Type
Availability Pricing
Traceable Two-Channel Digital Thermometer; 2 Wire Probes
Always in Stock
EW-94460-61 Mfr # 4307TR
COMPARE
Wire
$54.40USD / Each
Traceable Two-Channel Digital Thermometer; 2 Bottle Probes
Always in Stock
EW-94460-62 Mfr # 4306TR
COMPARE
Bottle
$90.04USD / Each