Traceable® Waterproof Food Thermometers
Waterproof pocket thermometer is ready for every temperature task
  • IP67-rated housing
  • One-second display updates
  • Position probe at any height using a unique holder
  • Flat design and lightweight for portability
  • Readings updated every second
  • One-key operation quickly displays MIN/MAX readings
  • °C/°F selectable
The waterproof food thermometer is engineered for professionals who require precise temperature measurements in food safety, food preparation, and laboratory environments. It handles a broad temperature range from –50 to 300°C (–58 to 572°F) and features a 3-1/2” long piercing stainless steel probe for use with liquids and semi-solids. Use the included metal clip for easy probe height adjustment—ideal for use with beakers, cylinders, or vats.
Lightweight, IP67-rated ABS body is safe for use in washdown areas, food labs, and in the field. Meters include a protective sleeve with clip, lanyard, and keychain attachment for easy portability and storage.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$37.00 - $49.60USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy
Availability Pricing
Traceable Waterproof Food Thermometer; ±1.5 °C Accuracy EW-98767-43 Mfr # 4420TR
COMPARE
±1.5°C (±3°F)
$37.00USD / Each
Traceable Waterproof Food Thermometer; ±0.4°C Accuracy at Tested Points
Always in Stock
EW-98767-44 Mfr # 4421TR
COMPARE
±0.4°C at tested points; otherwise ±1.0°C
$49.60USD / Each