Trajan Capillary Electrophoresis Tubing
Precision tubing for performance
  • Manufactured from precision bore tubing
  • hydrophobic and hydrophilic treatments enabling use with proteins, peptides, enzymes
$63.20 - $63.60USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 062803 Capillary Electrophoresis Tubing, Untreated, 363 um OD, 50 um ID, 1 m Length; 1/Ea EW-16006-93 Mfr # 062803
COMPARE
$63.60USD / Each
Trajan SGE 062823 Capillary Electrophoresis Tubing, Untreated, 363 um OD, 10 um ID, 1 m Length; 1/Ea EW-16006-95 Mfr # 062823
COMPARE
$63.20USD / Each