Trajan Fused Silica Capillary Tubing, Methyl Deactivated
Precision Tubing for Performance
  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.
$47.20 - $407.50USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 062442 Fused Silica Capillary Tubing, Methyl Deactivated, 285 um OD, 25 um ID, 2 m Length; 1/Ea EW-16006-32 Mfr # 062442
COMPARE
$47.30USD / Each
Trajan SGE 0624431 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 250 um ID, 2 m L; 1/Ea EW-16006-33 Mfr # 0624431
COMPARE
$47.20USD / Each
Trajan SGE 0624432 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 250 um ID, 5 m L; 1/Ea EW-16006-34 Mfr # 0624432
COMPARE
$95.00USD / Each
Trajan SGE 062445 Fused Silica Capillary Tubing, Methyl Deactivated, 220 um OD, 60 um ID, 2 m L; 1/Ea EW-16006-37 Mfr # 062445
COMPARE
$51.70USD / Each
Trajan SGE 06244501 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 125 um ID, 2 m L; 1/Ea EW-16006-39 Mfr # 06244501
COMPARE
$47.20USD / Each
Trajan SGE 06244502 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 75 um ID, 2 m L; 1/Ea EW-16006-40 Mfr # 06244502
COMPARE
$47.30USD / Each
Trajan SGE 06244503 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 50 um ID, 2 m L; 1/Ea EW-16006-41 Mfr # 06244503
COMPARE
$47.30USD / Each
Trajan SGE 0624455 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 100 um ID, 25 m L; 1/Ea EW-16006-42 Mfr # 0624455
COMPARE
$407.50USD / Each
Trajan SGE 0624459 Fused Silica Capillary Tubing, Methyl Deactivated, 310 um OD, 110 um ID, 2 m L; 1/Ea EW-16006-43 Mfr # 0624459
COMPARE
$47.60USD / Each
Trajan SGE 0624461 Fused Silica Capillary Tubing, Methyl Deactivated, 220 um OD, 150 um ID, 5 m L; 1/Ea EW-16006-46 Mfr # 0624461
COMPARE
$95.80USD / Each
Trajan SGE 0624465 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 150 um ID, 2 m L; 1/Ea EW-16006-48 Mfr # 0624465
COMPARE
$47.30USD / Each
Trajan SGE 0624469 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 220 um ID, 2 m L; 1/Ea EW-16006-49 Mfr # 0624469
COMPARE
$47.30USD / Each
Trajan SGE 062447 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 220 um ID, 5 m L; 1/Ea EW-16006-50 Mfr # 062447
COMPARE
$96.50USD / Each
Trajan SGE 0624471 Fused Silica Capillary Tubing, Methyl Deactivated, 430 um OD, 320 um ID, 5 m L; 1/Ea EW-16006-52 Mfr # 0624471
COMPARE
$95.30USD / Each
Trajan SGE 0624476 Fused Silica Capillary Tubing, Methyl Deactivated, 430 um OD, 320 um ID, 10 m L; 1/Ea EW-16006-56 Mfr # 0624476
COMPARE
$163.25USD / Each
Trajan SGE 0624479 Fused Silica Capillary Tubing, Methyl Deactivated, 680 um OD, 530 um ID, 2 m L; 1/Ea EW-16006-58 Mfr # 0624479
COMPARE
$47.30USD / Each
Showing 1- 16 of 16