Trajan Fused Silica Capillary Tubing, Non-Deactivated
Precision Tubing for Performance
  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.
$35.20 - $413.50USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 062454 Fused Silica Capillary Tubing, Non-Deactivated, 170 um OD, 110 um ID, 10 m L; 1/Ea EW-16006-62 Mfr # 062454
COMPARE
$133.25USD / Each
Trajan SGE 062456 Fused Silica Capillary Tubing, Non-Deactivated, 285 um OD, 5 um ID, 10 m L; 1/Ea EW-16006-63 Mfr # 062456
COMPARE
$132.50USD / Each
Trajan SGE 062458 Fused Silica Capillary Tubing, Non-Deactivated, 285 um OD, 10 um ID, 10 m L; 1/Ea EW-16006-65 Mfr # 062458
COMPARE
$133.25USD / Each
Trajan SGE 062460 Fused Silica Capillary Tubing, Non-Deactivated, 285 um OD, 25 um ID, 10 m L; 1/Ea EW-16006-66 Mfr # 062460
COMPARE
$133.25USD / Each
Trajan SGE 0624625 Fused Silica Capillary Tubing, Non-Deactivated, 140 um OD, 40 um ID, 10 m L; 1/Ea EW-16006-68 Mfr # 0624625
COMPARE
$134.25USD / Each
Trajan SGE 062463 Fused Silica Capillary Tubing, Non-Deactivated, 220 um OD, 50 um ID, 10 m L; 1/Ea EW-16006-69 Mfr # 062463
COMPARE
$133.25USD / Each
Trajan SGE 0624635 Fused Silica Capillary Tubing, Non-Deactivated, 150 um OD, 50 um ID, 10 m L; 1/Ea EW-16006-70 Mfr # 0624635
COMPARE
$133.25USD / Each
Trajan SGE 0624655 Fused Silica Capillary Tubing, Non-Deactivated, 220 um OD, 60 um ID, 10 m L; 1/Ea EW-16006-71 Mfr # 0624655
COMPARE
$133.25USD / Each
Trajan SGE 062466 Fused Silica Capillary Tubing, Non-Deactivated, 190 um OD, 75 um ID, 10 m L; 1/Ea EW-16006-72 Mfr # 062466
COMPARE
$133.25USD / Each
Trajan SGE 0624685 Fused Silica Capillary Tubing, Non-Deactivated, 200 um OD, 100 um ID, 10 m L; 1/Ea EW-16006-73 Mfr # 0624685
COMPARE
$133.25USD / Each
Trajan SGE 062470 Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 100 um ID, 25 m L; 1/Ea EW-16006-75 Mfr # 062470
COMPARE
$327.50USD / Each
Trajan SGE 062472 Fused Silica Capillary Tubing, Non-Deactivated, 220 um OD, 150 um ID, 10 m L; 1/Ea EW-16006-76 Mfr # 062472
COMPARE
$142.50USD / Each
Trajan SGE 062475 Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 220 um ID, 10 m L; 1/Ea EW-16006-79 Mfr # 062475
COMPARE
$133.25USD / Each
Trajan SGE 062478 Fused Silica Capillary Tubing, Non-Deactivated, 430 um OD, 320 um ID, 10 m L; 1/Ea EW-16006-81 Mfr # 062478
COMPARE
$133.25USD / Each
Trajan SGE 062479 Fused Silica Capillary Tubing, Non-Deactivated, 430 um OD, 320 um ID, 25 m L; 1/Ea EW-16006-82 Mfr # 062479
COMPARE
$324.50USD / Each
Trajan SGE 062482 Fused Silica Capillary Tubing, Non-Deactivated, 680 um OD, 530 um ID, 25 m L; 1/Ea EW-16006-84 Mfr # 062482
COMPARE
$413.50USD / Each
Trajan SGE 062711 Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 5 um ID, Custom EW-16006-88 Mfr # 062711
COMPARE
$35.20USD / Each
Trajan SGE 062712 Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 75 um ID, Custom EW-16006-89 Mfr # 062712
COMPARE
$35.60USD / Each
Showing 1- 18 of 18