Trajan Fused Silica Capillary Tubing, Phenyl Deactivated
Precision Tubing for Performance
  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.
$98.30 - $165.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 064062 Fused Silica Capillary Tubing, Phenyl Deactivated, 363 um OD, 250 um ID, 10 m L; 1/Ea EW-16007-03 Mfr # 064062
COMPARE
$165.25USD / Each
Trajan SGE 064102 Fused Silica Capillary Tubing, Phenyl Deactivated, 363 um OD, 220 um ID, 5 m L; 1/Ea EW-16007-04 Mfr # 064102
COMPARE
$98.30USD / Each