Trajan Glass-Lined Tubing
Combining Strength and Inertness
  • Can be used a straight tubing and can be machined or shaped
  • Strong acid and base resistance
  • Biocompatible, ideal of HPLC
  • Withstands high temperatures to 500°C
Addresses the problem of achieving an inert chromatographic system. Made by fusing borosilicate glass lining onto the internal surface of stainless steel tubing.
$42.70 - $259.25USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 082710 Glass Lined Tubing, 1/16 " OD, 0.3 mm ID, 180 cm Length; 1/Ea EW-16008-96 Mfr # 082710
COMPARE
$259.25USD / Each
Trajan SGE 082724 Glass Lined Tubing, 1/16 " OD, 0.7 mm ID, 90 cm Length; 1/Ea EW-16009-02 Mfr # 082724
COMPARE
$121.75USD / Each
Trajan SGE 0827352 Glass Lined Tubing, 1/16 " OD, 0.8 mm ID, 30 cm Length; 1/Ea EW-16009-05 Mfr # 0827352
COMPARE
$82.20USD / Each
Trajan SGE 0827353 Glass Lined Tubing, 1/16 " OD, 0.8 mm ID, 60 cm Length; 1/Ea EW-16009-06 Mfr # 0827353
COMPARE
$161.50USD / Each
Trajan SGE 082737 Glass Lined Tubing, 1/8 " OD, 1.0 mm ID, 30 cm Length; 1/Ea EW-16009-07 Mfr # 082737
COMPARE
$42.70USD / Each
Trajan SGE 082767 Glass Lined Tubing, 1/4 " OD, 4.0 mm ID, 30 cm Length; 1/Ea EW-16009-11 Mfr # 082767
COMPARE
$70.60USD / Each
Trajan SGE 082768 Glass Lined Tubing, 1/4 " OD, 4.0 mm ID, 60 cm Length; 1/Ea EW-16009-12 Mfr # 082768
COMPARE
$140.50USD / Each
Trajan SGE 08277028 Glass Lined Tubing, 1/2 " OD, 9.50 mm ID, 60 cm Length; 1/Ea EW-16009-14 Mfr # 08277028
COMPARE
$153.00USD / Each
Showing 1- 8 of 8