Trajan HPLC EASYLok
Quick and Easy HPLC connections
  • Double ended ferrule seals at two points
  • Knurled to make finger tightening easy
EasyLok™ fittings are composed of a knurled stainless steel nut and a double ended PEEK ferrule. The PEEK ferrule simply slides over any 1/16” OD tubing to its required position, while the nut is finger tightened. Unlike stainless steel, the PEEK ferrule will not crush the tubing and can be easily readjusted for quick column changes.
$53.60 - $76.80USD / Pkg of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 206102 EasyLOK Fitting with 2 fittings, 2 Ferrules; 2/Pk EW-15997-56 Mfr # 206102
COMPARE
$53.60USD / Pkg of 2
Trajan SGE 206160 PEEK™ Double Ended Ferrules; 10/Pk EW-15997-57 Mfr # 206160
COMPARE
$76.80USD / Pkg of 10