Trajan PEEKsil Tubing
Fused silica lined PEEK
  • Inert smooth flow path to minimize band broadening
  • Precise inner bore diameter
  • Flexible and robust
  • Color coded for easy identification of internal diameter
Compatible with most organic solvents, reduces dispersion and loss of efficiency, sensitivty, and resolution
$58.40 - $162.25USD / Pkg of 5
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 0624205 PEEKsil Tubing, 1/16" OD, 0.2 mm ID, 150 mm Length; 5/Pk EW-16005-61 Mfr # 0624205
COMPARE
$157.50USD / Pkg of 5
Trajan SGE 0624214 PEEKsil Tubing, 1/16" OD, 0.3 mm ID, 50 mm Length; 5/Pk EW-16005-62 Mfr # 0624214
COMPARE
$148.00USD / Pkg of 5
Trajan SGE 0624215 PEEKsil Tubing, 1/16" OD, 0.3 mm ID, 100 mm Length; 5/Pk EW-16005-63 Mfr # 0624215
COMPARE
$148.25USD / Pkg of 5
Trajan SGE 0624216 PEEKsil Tubing, 1/16" OD, 0.3 mm ID, 150 mm Length; 5/Pk EW-16005-64 Mfr # 0624216
COMPARE
$157.50USD / Pkg of 5
Trajan SGE 0624217 PEEKsil Tubing, 1/16" OD, 0.3 mm ID, 200 mm Length; 5/Pk EW-16005-65 Mfr # 0624217
COMPARE
$162.25USD / Pkg of 5
Trajan SGE 0624225 PEEKsil Tubing, 1/16" OD, 0.025 mm ID, 50 mm Length; 5/Pk EW-16005-67 Mfr # 0624225
COMPARE
$137.25USD / Pkg of 5
Trajan SGE 0624229 PEEKsil Tubing, 1/16" OD, 0.025 mm ID, 500 mm Length; 2/Pk EW-16005-71 Mfr # 0624229
COMPARE
$91.30USD / Pkg of 2
Trajan SGE 0624230 PEEKsil Tubing, 1/16" OD, 0.15 mm ID, 50 mm Length; 5/Pk EW-16005-72 Mfr # 0624230
COMPARE
$137.25USD / Pkg of 5
Trajan SGE 0624231 PEEKsil Tubing, 1/16" OD, 0.15 mm ID, 100 mm Length; 5/Pk EW-16005-73 Mfr # 0624231
COMPARE
$141.50USD / Pkg of 5
Trajan SGE 0624232 PEEKsil Tubing, 1/16" OD, 0.15 mm ID, 150 mm Length; 5/Pk EW-16005-74 Mfr # 0624232
COMPARE
$150.50USD / Pkg of 5
Trajan SGE 0624250 PEEKsil Tubing, 1/16" OD, 0.05 mm ID, 500 mm Length; 2/Pk EW-16005-82 Mfr # 0624250
COMPARE
$88.80USD / Pkg of 2
Trajan SGE 0624251 PEEKsil Tubing, 1/16" OD, 0.05 mm ID, 50 mm Length; 5/Pk EW-16005-83 Mfr # 0624251
COMPARE
$130.50USD / Pkg of 5
Trajan SGE 0624253 PEEKsil Tubing, 1/16" OD, 0.05 mm ID, 200 mm Length; 5/Pk EW-16005-85 Mfr # 0624253
COMPARE
$158.25USD / Pkg of 5
Trajan SGE 0624275 PEEKsil Tubing, 1/32" OD, 0.075 mm ID, 250 mm Length; 2/Pk EW-16005-93 Mfr # 0624275
COMPARE
$66.80USD / Pkg of 2
Trajan SGE 0624291 PEEKsil Tubing, 1/16" OD, 0.075 mm ID, 100 mm Length; 5/Pk EW-16005-96 Mfr # 0624291
COMPARE
$138.75USD / Pkg of 5
Trajan SGE 0624292 PEEKsil Tubing, 1/16" OD, 0.075 mm ID, 150 mm Length; 5/Pk EW-16005-97 Mfr # 0624292
COMPARE
Call For Price
Trajan SGE 0624294 PEEKsil Tubing, 1/16" OD, 0.075 mm ID, 500 mm Length; 2/Pk EW-16005-99 Mfr # 0624294
COMPARE
$88.80USD / Pkg of 2
Trajan SGE 0624301 PEEKsil Tubing, 1/16" OD, 0.1 mm ID, 50 mm Length; 5/Pk EW-16006-01 Mfr # 0624301
COMPARE
$145.50USD / Pkg of 5
Trajan SGE 0624303 PEEKsil Tubing, 1/16" OD, 0.1 mm ID, 200 mm Length; 5/Pk EW-16006-03 Mfr # 0624303
COMPARE
$158.25USD / Pkg of 5
Trajan SGE 0624313 PEEKsil Tubing, 1/32" OD, 0.1 mm ID, 150 mm Length; 2/Pk EW-16006-07 Mfr # 0624313
COMPARE
$59.90USD / Pkg of 2
Showing 1 - 20 of 28