Trajan ProteCol Fittings and Unions
Perfect connections, every time
  • Zero dead colume for no peak dispersion
  • Available in stainless steel or PEEK
  • Reusable PEEK ferrules, with replacements available
Allows for connection of any combination of 1/32" PEEKsil tubing, 1/16" PEEKsil tubing, 0.36mm fused silica tubing
$58.70 - $94.00USD / Pkg of 5
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 213007 ProteCol™ PEEK Ferrules, 1/16" to 1/32"; 5/Pk EW-15997-66 Mfr # 213007
COMPARE
$58.70USD / Pkg of 5
Trajan SGE 222017 ProteCol™ PEEK Fingertight Fittings, 1/32" to 6-40UNF Thread; 5/Pk EW-15997-74 Mfr # 222017
COMPARE
$94.00USD / Pkg of 5