Trajan SilTite Replacement Fittings
Available in two material types: stainless steel and Glass Lined Tubing
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
$111.00 - $218.75USD / Pkg of 10
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 073470 Replacement SilTite™ Mini Union Ferrules, Compatible with 0.1 - 0.25 mm ID Columns; 10/Pk EW-16008-46 Mfr # 073470
COMPARE
$111.00USD / Pkg of 10
Trajan SGE 073471 Replacement SilTite™ Mini Union Ferrules, Compatible with 0.32 mm ID Columns; 10/Pk EW-16008-47 Mfr # 073471
COMPARE
$111.00USD / Pkg of 10
Trajan SGE 073472 Replacement SilTite™ Mini Union Ferrules, Compatible with 0.45 - 0.53 mm ID Columns; 10/Pk EW-16008-48 Mfr # 073472
COMPARE
$111.50USD / Pkg of 10
Trajan SGE 073473 Replacement SilTite™ Mini Union Ferrules, for 1/32" Columns; 10/Pk EW-16008-49 Mfr # 073473
COMPARE
$111.50USD / Pkg of 10
Trajan SGE 073566 Replacement SilTite™ Micro Union Ferrules, for 0.25 and 0.25 mm Columns/Fused Silica; 10/Pk EW-16008-62 Mfr # 073566
COMPARE
$217.75USD / Pkg of 10
Trajan SGE 073567 Replacement SilTite™ Micro Union Ferrules, for 0.25 and 0.32 mm Columns/Fused Silica;10/Pk EW-16008-63 Mfr # 073567
COMPARE
$217.75USD / Pkg of 10
Trajan SGE 073568 Replacement SilTite™ Micro Union Ferrules, for 0.25 and 0.53 mm Columns/Fused Silica;10/Pk EW-16008-64 Mfr # 073568
COMPARE
$218.25USD / Pkg of 10
Trajan SGE 073570 Replacement SilTite™ Micro Union Ferrules, for 0.32 and 0.53 mm Columns/Fused Silica;10/Pk EW-16008-66 Mfr # 073570
COMPARE
$217.25USD / Pkg of 10
Trajan SGE 073571 Replacement SilTite™ Micro Union Ferrules, for 0.53 and 0.53 mm Columns/Fused Silica;10/Pk EW-16008-67 Mfr # 073571
COMPARE
$218.75USD / Pkg of 10
Showing 1- 9 of 9