TriForest Square/Octagonal Media Bottles, PC
Withstand multiple autoclave cycles
  • Polycarbonate (PC) is a safe and shatterproof alternative to glass
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom in an ISO 9001 compliant facility
  • Keep samples secure—leakproof seals included
  • Easily complete fill with molded-in graduations
Store buffers, reagents, media, and other critical fluids in these sturdy, clear bottles. The square shape saves shelf space by allowing bottles to be placed closer together.
Bottles come separated into packs by bags, and a case is double bagged to maintain cleanliness.
Autoclavable
$311.85 - $702.00USD / Case of 192
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Description
Availability Pricing
TriForest BPC0060 Square Autoclavable Media Bottle, 60 mL, PC, 24 per pack, 192/CS
Buy More Save More
EW-06542-20 Mfr # BPC0060
COMPARE
60 Square/Octagonal Media Bottle, 60 mL; 192/Cs
$503.00USD / Case of 192
TriForest BPC0125 Square Autoclavable Media Bottle, 125 mL, PC, 24 per pack, 144/CS
Buy More Save More
EW-06542-21 Mfr # BPC0125
COMPARE
125 Square/Octagonal Media Bottle, 125 mL; 144/Cs
$702.00USD / Case of 144
TriForest BPC0250 Square Autoclavable Media Bottle, 250 mL, PC, 24 per pack, 96/CS
Buy More Save More
EW-06542-22 Mfr # BPC0250
COMPARE
250 Square/Octagonal Media Bottle, 250 mL; 96/Cs
$597.00USD / Case of 96
TriForest BPC0500 Square Autoclavable Media Bottle, 500 mL, PC, 12 per pack, 72/CS
Buy More Save More
EW-06542-23 Mfr # BPC0500
COMPARE
500 Square/Octagonal Media Bottle, 500 mL; 72/Cs
$538.00USD / Case of 72
TriForest BPC1000 Square Autoclavable Media Bottle, 1 L, PC, 12 per pack, 12/Cs
Buy More Save More
EW-06542-24 Mfr # BPC1000
COMPARE
1000 Square Autoclavable Media Bottle, 1 L, PC, 12 per pack, 12/Cs
$311.85USD / Case of 12
TriForest BPC2000 Square Autoclavable Media Bottle, 2 L, PC, 6 per pack, 24/CS
Buy More Save More
EW-06542-25 Mfr # BPC2000
COMPARE
2000 Square/Octagonal Media Bottle, 2000 mL, 24/Cs
$340.50USD / Case of 24