Troemner Calibration Mass Tweezers
Forceps / Tweezers are designed for proper handling of weights.
Stainless steel forceps are available in 5” and 8” sizes and are designed in a smooth tip, curved tip, straight grip tip, and a 22° grip tip. Stainless steel forceps tips are made of PVC.
Economical polycarbonate forceps for weights 1 mg through 200 g.
$26.90 - $101.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Description
Availability Pricing
Troemner WA056 Polycarbonate Tweezer for Weights 1mg, Through 200G
Always in Stock
EW-01027-88 Mfr # WA056
COMPARE
Polycarbonate 4 Troemner WA056 Polycarbonate Tweezer for Weights 1mg, Through 200G
$26.90USD / Each
Troemner WA160 5 and Quot; S/S Tweezer with 22 and #176; Grip Tip for Weights 1g Through 200g EW-01027-89 Mfr # WA160
COMPARE
Stainless steel 5 Troemner WA160 5 and Quot; S/S Tweezer with 22 and #176; Grip Tip for Weights 1g Through 200g
$62.20USD / Each
Troemner WA220 5" S/S Tweezer Grip Tip for Weights 1g Through 200g EW-01027-90 Mfr # WA220
COMPARE
Stainless steel 5 Troemner WA220 5" S/S Tweezer Grip Tip for Weights 1g Through 200g
$62.20USD / Each
Troemner WA208 5" S/S Tweezer Smooth Tip for mg, Weights EW-01027-91 Mfr # WA208
COMPARE
Stainless steel 5 Troemner WA208 5" S/S Tweezer Smooth Tip for mg, Weights
$62.20USD / Each
Troemner WA223 5" S/S Tweezer Curved Tip for mg, Weights EW-01027-92 Mfr # WA223
COMPARE
Stainless steel 5 Troemner WA223 5" S/S Tweezer Curved Tip for mg, Weights
$62.20USD / Each
Troemner WA212 8" S/S Tweezer with 22 and #176; Grip Tip for Weights 1g Through 200g EW-01027-93 Mfr # WA212
COMPARE
Stainless steel 8 Troemner WA212 8" S/S Tweezer with 22 and #176; Grip Tip for Weights 1g Through 200g
$101.00USD / Each
Troemner WA228 8" S/S Tweezer Grip Tip for Weights 1g Through 200g EW-01027-94 Mfr # WA228
COMPARE
Stainless steel 8 Troemner WA228 8" S/S Tweezer Grip Tip for Weights 1g Through 200g
$101.00USD / Each
Showing 1- 7 of 7