TSI Alnor Rotating Vane Anemometers
Versatile handheld or mountable design for quick air velocity sampling or long term monitoring
  • Measure air velocities of 50 to 6000 ft/min (0.25–30 m/s) to cover a broad range of applications
  • Digital display for easy reading
The Alnor® Rotating Vane Anemometers measure air velocity, air volume, and temperature in imperial or metric units. The 4-inch (100mm) head automatically averages velocity and volume readings, making them essential tools for measuring unevenly distributed or fluctuating flows.
$812.00 - $1,280.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Storage (samples) Max Air Velocity (ft/min) Description
Availability Pricing
TSI RVA801 Hand-held vane anemometer EW-10374-86 Mfr # RVA801
COMPARE
None 6000 Hand-held vane anemometer
$812.00USD / Each
TSI RVA501 Data Logging Vane Anemometer Hand Held EW-10508-74 Mfr # RVA501
COMPARE
12,700 samples and 100 test IDs 6000 Data Logging Vane Anemometer Hand Held
$1,280.00USD / Each