Tygon® 2475 Plasticizer Free Thermoplastic Tubing
Resistant to aggressive acids, bases, and solvents
  • Low gas permeation
  • Smooth inner surface inhibits particle entrapment
  • Free from added plasticizers or oils
  • Meets USP Class VI criteria
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Due to the absence of plasticizers, the Tygon® 2475 high-purity thermoplastic tubing can stand up to aggressive acids, bases, and solvents that would react with or extract plasticizer from a standard PVC tube. The clear tubing allows you to visually inspect the fluid path to ensure consistency of flow. Designed with lower gas permeation than standard silicone tubing, sensitive fluids are protected from moisture loss and oxidation.
The surface smoothness of the tubing helps reduce potential microbial buildup, critical in bioprocess applications. The tubing also features a very low absorption of aqueous substances, minimizing the risk of fluid alteration in single or repeat use applications.
Applications include aseptic filling and dispensing systems, diagnostic equipment, nuclear equipment, lab analytical instrumentation, cosmetic production, and cell and tissue culture transport.
$102.00 - $640.00USD / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon 2475 Tubing, 1/16" ID x 3/16" OD; 50 Ft EW-50113-72 Mfr # ACG00003
COMPARE
1/16 3/16 1/16
$102.00USD / Pkg of 1
Tygon 2475 Tubing, 1/8" ID x 1/4" OD; 50 Ft EW-50113-73 Mfr # ACG00007
COMPARE
1/8 1/4 1/16
$157.50USD / Pkg of 1
Tygon 2475 Tubing, 3/16" ID x 5/16" OD; 50 Ft EW-50113-74 Mfr # ACG00012
COMPARE
3/16 5/16 1/16
$173.25USD / Pkg of 1
Tygon 2475 Tubing, 1/4" ID x 3/8" OD; 50 Ft EW-50113-75 Mfr # ACG00017
COMPARE
1/4 3/8 1/16
$183.75USD / Pkg of 1
Tygon 2475 Tubing, 5/16" ID x 7/16" OD; 50 Ft EW-50113-76 Mfr # ACG00022
COMPARE
5/16 7/16 1/16
$215.25USD / Pkg of 1
Tygon 2475 Tubing, 3/8" ID x 1/2" OD; 50 Ft EW-50113-77 Mfr # ACG00027
COMPARE
3/8 1/2 1/16
$236.25USD / Pkg of 1
Tygon 2475 Tubing, 1/2" ID x 3/4" OD; 50 Ft EW-50113-78 Mfr # ACG00038
COMPARE
1/2 3/4 1/8
$520.00USD / Pkg of 1
Tygon 2475 Tubing, 5/8" ID x 7/8" OD; 50 Ft EW-50113-79 Mfr # ACG00046
COMPARE
5/8 7/8 1/8
$598.00USD / Pkg of 1
Tygon 2475 Tubing, 3/4" ID x 1" OD; 50 Ft EW-50113-80 Mfr # ACG00053
COMPARE
3/4 1 1/8
$640.00USD / Pkg of 1
Tygon 2475 Tubing, 1" ID x 1- 3/8" OD; 25 Ft EW-50113-81 Mfr # ACG42064
COMPARE
1 1 3/8 3/16
$552.00USD / Pkg of 1
Showing 1- 10 of 10