Tygon® S3™ B-44-4X Food and Beverage Tubing
Smooth, nonporous bore inhibits particle entrapment
  • Sanitary fluid path minimizes potential for bacterial growth
  • Lightweight, flexible, and easy to handle
  • Withstands harsh alkaline cleaners and sanitizers
  • Phthalate and BPA free
  • FDA and 3A compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Tygon® S3™ B-44-4X tubing is specifically designed for food and beverage applications. The smooth, nonporous bore promotes easy filling, draining, and transferring of fluids without trapping any particles that can cause bacterial growth. This lightweight and flexible tubing bends to accommodate corners and obstructions and requires minimal fittings for quick and easy installation. Clear tubing allows you to visually monitor the flow, so you can control or adjust the flow during the process.
Complies with applicable FDA Food Additive Regulations, meets 3A Sanitary Standard 20-27, and is not intentionally made or manufactured with animal-derived materials.
$11.50 - $593.00USD / Pkg of 1
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/32" ID x 3/32" OD; 50 Ft EW-22050-69 Mfr # AAA00001
COMPARE
1/32 3/32 1/32
$11.50USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/16" ID x 1/8" OD; 50 Ft EW-22050-70 Mfr # AAA00002
COMPARE
1/16 1/8 1/32
$18.60USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 3/32" ID x 5/32" OD; 50 Ft EW-22050-71 Mfr # AAA00004
COMPARE
3/32 5/32 1/32
$24.40USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/8" ID x 3/16" OD; 50 Ft EW-22050-72 Mfr # AAA00006
COMPARE
1/8 3/16 1/32
$25.50USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/8" ID x 1/4" OD; 50 Ft
Always in Stock
EW-50106-77 Mfr # AAA00007
COMPARE
1/8 1/4 1/16
$70.40USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 3/16" ID x 5/16" OD; 50 Ft EW-50106-78 Mfr # AAA00012
COMPARE
3/16 5/16 1/16
$97.70USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/4" ID x 3/8" OD; 50 Ft EW-50106-79 Mfr # AAA00017
COMPARE
1/4 3/8 1/16
$120.75USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/4" ID x 7/16" OD; 50 Ft EW-50106-80 Mfr # AAA00018
COMPARE
1/4 7/16 3/32
$194.25USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 5/16" ID x 7/16" OD; 50 Ft EW-50106-81 Mfr # AAA00022
COMPARE
5/16 7/16 1/16
$141.75USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 5/16" ID x 1/2" OD; 50 Ft EW-50106-82 Mfr # AAA00023
COMPARE
5/16 1/2 3/32
$204.75USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 3/8" ID x 1/2" OD; 50 Ft EW-50106-83 Mfr # AAA00027
COMPARE
3/8 1/2 1/16
$162.75USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 3/8" ID x 9/16" OD; 50 Ft EW-50106-84 Mfr # AAA00028
COMPARE
3/8 9/16 3/32
$210.00USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1/2" ID x 3/4" OD; 50 Ft EW-50106-85 Mfr # AAA00038
COMPARE
1/2 3/4 1/8
$373.00USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 5/8" ID x 7/8" OD; 50 Ft EW-50106-86 Mfr # AAA00046
COMPARE
5/8 7/8 1/8
$420.00USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 3/4" ID x 1" OD; 50 Ft EW-50106-87 Mfr # AAA00053
COMPARE
3/4 1 1/8
$478.00USD / Pkg of 1
Tygon S3™ B-44-4X Food and Beverage Tubing, 1" ID x 1-1/4" OD; 50 Ft EW-50106-88 Mfr # AAA00062
COMPARE
1 1 1/4 1/8
$593.00USD / Pkg of 1
Showing 1- 16 of 16