Tygon® S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing
Excellent for dispensing food and beverages at elevated pressures
  • All layers compliant with FDA and EU food contact regulations
  • Smooth, nonporous bore
  • Lightweight and easy to handle
  • Clear wall provides visual monitoring of flow
  • Excellent chemical resistance
  • Phthalate and BPA free
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Reinforced to handle higher pressures, Tygon S3 B-44 5X IB pressure tubing is perfect for dispensing thick fluids such as ice cream, smoothies, and shakes. The smooth, nonporous bore promotes easy filling, draining, and transferring of fluids without trapping any particles that can cause bacterial growth.
The nonwetting properties of the tubing provide complete drainage and allow simple flush cleaning. Lightweight and flexible, the tubing bends to accommodate corners and obstructions and requires minimal fittings for quick and easy installation. A clear wall enables visual monitoring of the flow, so you can control or adjust the flow during the process.
$199.50 - $2,565.00USD / Pkg of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 1/4" ID x 7/16" OD; 50 Ft EW-50122-40 Mfr # AMH00018
COMPARE
50 1/4 7/16
$199.50USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 3/8" ID x 5/8" OD; 50 Ft EW-50122-41 Mfr # AMH00029
COMPARE
50 3/8 5/8
$310.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 1/2" ID x 3/4" OD; 50 Ft EW-50122-42 Mfr # AMH00038
COMPARE
50 1/2 3/4
$409.50USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 5/8" ID x 7/8" OD; 50 Ft EW-50122-43 Mfr # AMH00046
COMPARE
50 5/8 7/8
$515.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 3/4" ID x  1" OD; 50 Ft EW-50122-44 Mfr # AMH00053
COMPARE
50 3/4 1
$562.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 1" ID x 1-3/8" OD; 50 Ft EW-50122-45 Mfr # AMH00064
COMPARE
50 1 1 3/8
$957.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 1-1/4" ID x 1-5/8" OD; 50 Ft EW-50122-46 Mfr # AMH00070
COMPARE
50 1 1/4 1 5/8
$1,460.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 1-1/2" ID x  2" OD; 50 Ft EW-50122-47 Mfr # AMH00074
COMPARE
50 1 1/2 2
$2,125.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 1-1/2" ID x  2" OD; 20 Ft EW-50122-48 Mfr # AMH05074
COMPARE
20 1 1/2 2
$1,345.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 2" ID x 2-3/4" OD; 20 Ft EW-50122-49 Mfr # AMH05079
COMPARE
20 2 2 3/4
$1,610.00USD / Pkg of 1
Tygon S3™ B-44-5X IB Reinforced Food and Beverage Dispensing Tubing, 3" ID x 3-3/4" OD; 20 Ft EW-50122-50 Mfr # AMH05086
COMPARE
20 3 3 3/4
$2,565.00USD / Pkg of 1
Showing 1- 11 of 11