Tygon® S3™ E-3603 Metric Vacuum Tubing
Formulated to resist flex-fatigue and abrasion
  • Bio-based formulation reduces environmental impact
  • Lot-to-lot consistency for reproducible results
  • Easily slips over fittings and grips securely
  • Contains no BPA or phthalates
  • Meets the requirements of the USP, Biological Test for Plastics, Class VI
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Able to handle most organic chemicals found in the lab, the clear and flexible Tygon® S3™ E-3603 is non-oxidizing and non-contaminating. The tubing is less permeable than rubber tubing and features a glassy-smooth inner bore which prevents buildup and facilitates easy cleaning.
Designed to resist flex-fatigue and abrasions, the tubing is constructed with extra-heavy walls which can withstand a full vacuum at room temperature. It can be sterilized through conventional autoclave methods and ethylene oxide.
Applications include general laboratory, analytical instruments, and peristaltic and vacuum pumps. Ideal for condensers, desiccators, gas lines, and drain lines.
$41.40 - $230.50USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
Tygon E-3603 Metric Tubing, 2 mm ID x 4 mm OD; 15 Meters EW-50143-57 Mfr # ACF1S1502
COMPARE
2 4 1
$41.40USD / Pkg of 1
Tygon E-3603 Metric Tubing, 3 mm ID x 5 mm OD; 15 Meters EW-50143-58 Mfr # ACF1S1517
COMPARE
3 5 1
$53.60USD / Pkg of 1
Tygon E-3603 Metric Tubing, 4 mm ID x 6 mm OD; 15 Meters EW-50143-59 Mfr # ACF1S1518
COMPARE
4 6 1
$65.40USD / Pkg of 1
Tygon E-3603 Metric Tubing, 5 mm ID x 8 mm OD; 15 Meters EW-50143-60 Mfr # ACF1S1503
COMPARE
5 8 1.5
$125.00USD / Pkg of 1
Tygon E-3603 Metric Tubing, 6 mm ID x 9 mm OD; 15 Meters EW-50143-61 Mfr # ACF1S1504
COMPARE
6 9 1.5
$139.75USD / Pkg of 1
Tygon E-3603 Metric Tubing, 8 mm ID x 12 mm OD; 15 Meters EW-50143-62 Mfr # ACF1S1506
COMPARE
8 12 2
$230.50USD / Pkg of 1