Vacuum Grease

Stiff, nonmelting silicone lubricant maintains its consistency from –40 to 500°F
  • Dimethyle siloxane/silicon dioxide
  • Comes in 5.3 oz tube
Vacuum grease is a stiff, nonmelting silicone lubricant that maintains its consistency from –40 to 500°F. Grease is nonoxidizing and nongumming and has excellent resistance to water and chemicals.
$79.10 - $264.75USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material
Availability Pricing
DuPont MOLYKOTE® High Vacuum Grease, 5.3 oz
Always in Stock
EW-79751-30 Mfr # DC976VF
COMPARE
Silicone compound
$79.10USD / Each
RT-15 Vacuum Grease, Propene / PTFE, 392 °F Max Temp; 3.5 Oz. EW-79751-36 Mfr # FOMBLIN RT-15 GREASE
COMPARE
Propene/PTFE
$264.75USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat