Vaisala HMT 120 Series Humidity / Temperature / Dew Point Transmitter
Features relative humidity with the option of dew point, and temperature outputs
  • Probe can be easily removed and replaced without having to adjust the transmitter
  • Two-wire, loop-powered output configurations
  • Fixed and remote probe models
  • Optional LCD display
  • Vaisala HUMICAP® sensor
  • Different output scalings
  • Compatible with handheld HM70 and HMI41 for one-point calibration
  • IP65 (NEMA 4) protective housing
Vaisala HMT120 series transmitters are designed for monitoring humidity and temperature in demanding environments. Typical applications include stability rooms, HVAC, livestock, indoor swimming pools, and outdoor applications. The transmitters can be wall mounted and come with either an integral or remote probe, and with or without a display. The smooth surface of the enclosure makes it easy to clean and tolerates purifying agents. Remote probe is made of chrome coated aluminum and is ideal for high-temperature applications or where space is limited.
Note: Other options may be available — call for more information.
$663.00 - $1,335.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max RH Range (%) Max Temperature (° F)
Availability Pricing
Vaisala HMT120 RH/ Only Transmitter with No Display, Wall Mount and 4-20mA Output EW-37801-40 Mfr # HMT120-HA0A1A22A1A0Z
COMPARE
None 100 176
$663.00USD / Each
Vaisala HMT120 RH/ Only Transmitter with Display, Wall Mount and 4-20mA Output EW-37801-41 Mfr # HMT120-HA1A1A22A1A0Z
COMPARE
LCD 100 176
$778.00USD / Each
Vaisala HMT120 RH//Temperature Transmitter; No Display, Wall-Mt, 4-20Ma EW-37801-46 Mfr # HMT120-KA0A1B22A1A0Z
COMPARE
None 100 176
$824.00USD / Each
Vaisala HMT120 RH/ and Temperature Transmitter with Display, Wall Mount and 4-20mA Output EW-37801-47 Mfr # HMT120-KA1A1B22A1A0Z
COMPARE
LCD 100 176
$939.00USD / Each
Vaisala HMT120 RH//Temperature Transmitter; No Display, 3M Probe, 4-20Ma EW-37801-48 Mfr # HMT120-KB0A1B22A1A0Z
COMPARE
None 100 176
$873.00USD / Each
Vaisala HMT120 RH//Temp/Dew-Pt Transmitter; Display, Wall-Mt, 4-20Ma EW-37801-55 Mfr # HMT120-KA1A2B22A1A0Z
COMPARE
LCD 100 176
$939.00USD / Each
Vaisala HMT120 RH//Temp/Dew-Pt Transmitter; No Display, 5M Probe, 4-20Ma EW-37801-56 Mfr # HMT120-KC0A2B22A1A0Z
COMPARE
None 100 176
$899.00USD / Each
Vaisala HMT120 RH//Temp/Dew-Pt Transmitter; Display, 5M Probe, 4-20Ma EW-37801-57 Mfr # HMT120-KC1A2B22A1A0Z
COMPARE
LCD 100 176
$1,335.00USD / Each
Showing 1- 8 of 8