Vaisala Indigo80 Handheld Meters
Industrial-grade handheld indicator for field sampling, data analysis, and diagnostics of multiple parameters
  • Simultaneously accepts up to two Indigo-compatible probes or transmitters
  • Multilingual, menu-based user interface available in 10 languages
  • Logs up to 5.5 million real-time data values
  • Durable aluminum body resists dust and most chemicals
Use these high-accuracy portable meters to spot check, monitor, configure, and troubleshoot to ensure your process is running correctly and efficiently. Robust and reliable Indigo80 handheld meters accommodate up to two Indigo-compatible probes or transmitters. Measure humidity, temperature, dew point, carbon dioxide, hydrogen peroxide vapor, or moisture in oil by connecting appropriate probes/transmitters to the meter.
Multilingual, intuitive, menu-based user interface is easy to navigate and use. Easily view live measurement data both numerically and graphically on the 2.7" sunlight readable TFT LCD color display; backlight with automatic brightness control makes it easy to view readings in dimly lit areas. Meters log up to 5.5 million real-time data values for analysis. Connect the meters to free Vaisala Insight software to access logged data and configuration functionality. Use the USB-C interface for data uploads and battery charging. A rechargeable lithium-ion battery provides a typical operation time of 10 hours; connect to AC power for long-term logging (order separately).
$2,975.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Data Logging Display Type Description
Availability Pricing
Vaisala Indigo80 Handheld Meter with Flat Cable for GMP251 Probe Connection EW-37805-38 Mfr # INIDGO80 D1A2A1N0
COMPARE
Yes Transflective TFT LCD color display Indigo80 Handheld Meter with Flat Cable for GMP251 Probe Connection
$2,975.00USD / Each
Vaisala Indigo80 Handheld Meter EW-37805-39 Mfr # INDIGO80 D1A1A1N0
COMPARE
Yes Transflective TFT LCD color display Indigo80 Handheld Meter
$2,975.00USD / Each