Vanguard Safety A63C1 Hybrid EVA Industrial Gloves, 4 mil
Offers Secure grip and increased dexterity
  • Recyclable
  • Powder-free
  • Complies with 21 CFR parts 170-199 for use in contact with food
  • Ambidextrous
These disposable hybrid EVA industrial gloves are an ideal alternative for short-term use and applications that require frequent glove changes. The 4-mil thick clear gloves are powder-free to reduce skin irritation, embossed for a secure grip, and latex free. Stretchy fit design makes getting the gloves on easy and offers increased dexterity. Ideal for use in food service, janitorial, beauty, and industrial industries.
$60.40 - $64.80USD / Case of 2000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Size
Availability Pricing
Vanguard Safety Hybrid EVA Industrial Gloves, 4 mil,  Small; 2000/CS EW-78906-40 Mfr # A63C11
COMPARE
9 Ethylene-vinyl acetate (EVA) Small
$60.40USD / Case of 2000
Vanguard Safety Hybrid EVA Industrial Gloves, 4 mil,  Medium; 2000/CS EW-78906-41 Mfr # A63C12
COMPARE
9 Ethylene-vinyl acetate (EVA) Medium
$64.80USD / Case of 2000
Vanguard Safety Hybrid EVA Industrial Gloves, 4 mil,  Large; 2000/CS EW-78906-42 Mfr # A63C13
COMPARE
9 Ethylene-vinyl acetate (EVA) Large
$60.40USD / Case of 2000
Vanguard Safety Hybrid EVA Industrial Gloves, 4 mil,  Extra-Large; 2000/CS EW-78906-43 Mfr # A63C14
COMPARE
9 Ethylene-vinyl acetate (EVA) X-Large
$60.40USD / Case of 2000