Vanguard Safety C11A2 Nitrile Unlined Gloves, 11 mil, 13" Cuff, Green
Maintain secure grip in dry, wet, and oily conditions
  • Diamond textured non-slip surface ensures secure grip
  • Straight cuff
  • Complies with FFDCA food handling requirements 21 CFR, Parts 170-199
These reusable nitrile unlined gloves offer you durable protection from grease, fats, and heavy-duty cleaning solutions. Diamond textured non-slip surface offers secure grip in dry, wet, and oily conditions. The 11-mil thick gloves are puncture and abrasion resistant. Ideal for use in food processing and manufacturing plants.
$279.25USD / Case of 144
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Size Thickness (mil)
Availability Pricing
Vanguard Safety Nitrile Unlined Gloves, 11 mil, 13" Cuff, Green, Medium; 144/CS EW-78906-66 Mfr # C11A28
COMPARE
Nitrile Medium 11
$279.25USD / Case of 144
Vanguard Safety Nitrile Unlined Gloves, 11 mil, 13" Cuff, Green, Large; 144/CS EW-78906-67 Mfr # C11A29
COMPARE
Nitrile Large 11
$279.25USD / Case of 144
Vanguard Safety Nitrile Unlined Gloves, 11 mil, 13" Cuff, Green, Extra-Large; 144/CS EW-78906-68 Mfr # C11A210
COMPARE
Nitrile Extra Large 11
$279.25USD / Case of 144