Versilon® ETFE Tubing
Durable ETFE tubing designed for mechanical strength and radiation-resistant performance
  • High mechanical strength and impact resistance provide durability in demanding environments
  • Resistant to radiation, abrasion, and weathering for long-term reliability
  • Good chemical resistance supports use with a wide range of fluids and solvents
  • Broad temperature capability up to ~300°F (148°C) for consistent performance across applications
Versilon® ETFE tubing is engineered for applications requiring a combination of mechanical durability, chemical resistance, and environmental stability. Offering improved strength and impact resistance compared to many fluoropolymers, it is well-suited for use in demanding conditions where abrasion, stress, or repeated handling are concerns. Its resistance to radiation and weathering makes it a reliable choice for specialized environments, while its chemical compatibility supports use with a wide range of solvents and process fluids. With a continuous service temperature of up to approximately 300°F (148°C), Versilon® ETFE tubing provides dependable performance in both industrial and laboratory applications, including components such as fittings, valves, and pump systems.
$110.25 - $737.00USD / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon ETFE Tubing, 1/50" ID x 1/16" OD, 50 ft
New Products
EW-48526-49 Mfr # TSTZ83-0062-021-50
COMPARE
1/50 1/16 1/50
$267.75USD / Pkg of 1
Versilon ETFE Tubing, 1/64" ID x 1/16" OD, 50 ft
New Products
EW-48526-50 Mfr # TSTZ83-0062-023-50
COMPARE
1/50 1/16 1/50
$283.50USD / Pkg of 1
Versilon ETFE Tubing, 3/32" ID x 1/8" OD, 50 ft
New Products
EW-48526-51 Mfr # TSTZ83-0125-016-50
COMPARE
3/32 1/8 1/64
$110.25USD / Pkg of 1
Versilon ETFE Tubing, 1/16" ID x 1/8" OD, 50 ft
New Products
EW-48526-52 Mfr # TSTZ83-0125-031-50
COMPARE
1/16 1/8 1/16
$373.00USD / Pkg of 1
Versilon ETFE Tubing, 3/16" ID x 1/4" OD, 50 ft
New Products
EW-48526-53 Mfr # TSTZ83-0250-031-50
COMPARE
3/16 1/4 1/16
$246.75USD / Pkg of 1
Versilon ETFE Tubing, 1/8" ID x 1/4" OD, 50 ft
New Products
EW-48526-54 Mfr # TSTZ83-0250-062-50
COMPARE
1/8 1/4 1/16
$541.00USD / Pkg of 1
Versilon ETFE Tubing, 15/64" ID x 5/16" OD, 50 ft
New Products
EW-48526-55 Mfr # TSTZ83-0312-040-50
COMPARE
15/64 5/16 1/25
$423.50USD / Pkg of 1
Versilon ETFE Tubing, 9/32" ID x 3/8" OD, 50 ft
New Products
EW-48526-56 Mfr # TSTZ83-0375-048-50
COMPARE
9/32 3/8 3/64
$441.00USD / Pkg of 1
Versilon ETFE Tubing, 3/8" ID x 1/2" OD, 50 ft
New Products
EW-48526-57 Mfr # TSTZ83-0500-062-50
COMPARE
3/8 1/2 1/16
$737.00USD / Pkg of 1
Showing 1- 9 of 9